Learn More
Thermo Scientific™ Orion™ AQUAfast AQ4500 IR and White Light Dual Source Waterproof Turbidity Meter, Turbidimeter and Calibration Kit PROMO
Shop All Thermo Scientific ProductsDescription
Log data into internal memory, or order AQ4CBL cable to send data to printer or download to a computer for easy time- and date-stamped results storage.
- U.S. EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg. 38351-38356
- Dual source to comply with EPA 180.1 and ISO 7027
- Nephelometric and ratiometric measurements with autoranging
- IP67 waterproof portable turbidity meter
- Readings in the range of 0 to 4000 NTU with a choice of units: NT, FTU, FNU, ASBC or EBC
Includes:
- AQ4500 meter
- Set of 5 USEPA approved primary calibration standards
- Seven glass vials
- User guide
- Carrying case
Specifications
Specifications
| Type | Turbidity Meter |
| Light Source | LED, Multiple |
| Display Type | LED |
| Automatic Range Selection | Yes |
| Description | Turbidimeter and Calibration Kit |
| Battery Life | 2500 hr. |
| EPA Approved Methods | 180.1 |
| Certifications/Compliance | US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7032 |
| ISO Accepted Methods | ISO 7027 |