Learn More
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Specifications
| Type | Ammonia |
| Test Reagent Formats | Ampoule |
| For Use With (Equipment) | Orion AQUAfast AQ4000 Colorimeter |
| Description | AQUAfast ammonia ULR Auto-ID ampoules, 30 pack |
| Includes | 30 Auto-ID Ampoules |
| Method | Salicylate |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 0.2 to 3 mg/L N |
| Quantity | 30 Ampoules |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.