Learn More
Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick
Shop All Thermo Scientific ProductsDescription
Use the Thermo Scientific™ Orion™ AquaMate Preprogrammed Methods on USB Memory Stick with over 260 preprogrammed methods for Orion™, CHEMetrics and Merck reagent chemistries on the Orion™ AquaMate 8000 UV-Vis and Orion™ AquaMate 7000 Vis spectrophotometers. Preprogrammed methods can be added, removed or modified on the Orion™ AquaMate spectrophotometers at any time.
- Compatible with the AquaMate 8000 UV-Vis and AquaMate 7000 Vis Spectrophotometers.
Specifications
Specifications
| Type | USB Memory Stick |
| For Use With (Equipment) | Orion AquaMate spectrophotometers |
| Includes | Over 260 preprogrammed methods compatible with several leading brands of reagents |