Learn More
Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes
Shop All Thermo Scientific ProductsDescription
Orion™ AquaPro™ Electrodes have polymer filled, double junction reference systems that are low maintenance. They have an extended life, fast response and clog resistant open junctions and are ideal for use in Tris, sulfides, proteins and biological media. The junction must be kept wet when not in use.
Solutions required
Electrode storage solution
- 910001 pH electrode storage solution, 475 mL bottle
pH buffers
- 910168 pH 1.68 buffer, NIST traceable, 475 mL
- 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
- 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
- 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
- 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
- 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
- 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
- 910112 pH 12.46 buffer, NIST traceable, 475 mL
- 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
- 900020 Cleaning solution A and C, 30 ml each. B and D, 60 mL each
Warranty period
- 12 months from date of purchase
Specifications
Specifications
| Description | Epoxy Body pH/ATC |
| Connector Types | Mini-DIN (Star) |
| Junction | Open |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | pH/ATC Combination Electrode |
| For Use With (Application) | General Purpose |
| For Use With (Equipment) | Orion Star Series Meters |
| Internal Reference | Ag/AgCl Double Junction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.