Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes

Catalog No. 13645154 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Mini-DIN (Star)
Material:
Epoxy Body
Glass Body
Type:
Combination pH Electrode
pH/ATC Combination Electrode
Tip Type:
Flat Surface
Standard
Toughened Bulb
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Material Type Tip Type
13-645-154 BNC (Waterproof) Epoxy Body Combination pH Electrode Standard
13-645-151 BNC (Waterproof) Glass Body Combination pH Electrode Toughened Bulb
13-644-966 Mini-DIN (Star) Epoxy Body pH/ATC Combination Electrode Standard
13-645-153 BNC (Waterproof) Epoxy Body Combination pH Electrode Flat Surface
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-645-154 Supplier Thermo Scientific™ Supplier No. 9156APWP
  • $783.50
    $736.65 / Each of 1

    Save $46.85

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measure the pH of viscous liquid samples quickly, and accurately, with clog-resistant Orion™ AquaPro™ pH electrodes.

Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
 

Orion™ AquaPro™ Electrodes have polymer filled, double junction reference systems that are low maintenance. They have an extended life, fast response and clog resistant open junctions and are ideal for use in Tris, sulfides, proteins and biological media. The junction must be kept wet when not in use.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 mL each

Warranty period

  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY

Specifications

Description Epoxy Body
Connector Types BNC (Waterproof)
Junction Open
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type Combination pH Electrode
For Use With (Application) General Purpose, rugged use, lab or field use
For Use With (Equipment) Standard pH Meter
Internal Reference Ag/AgCl Double Junction
pH Relative Accuracy ±0.02
Storage Requirements Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Temperature (English) Operating 32°F to 140°F
Tip Type Standard
Length (English) 4.72 in.
Length (English) Cable 39 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.