Learn More
Description
Save time and enhance data with the Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler, which automatically performs analyses, calibrations and standardization procedures.
Efficient and durable:
- Improves data quality by using up to five rinse stations
- Constructed of durable, corrosion-resistant, sealed electronics chassis and coated metal parts to protect against harshest chemical environments
- Compatible with Orion™ Star™ Plus meters and 960 Titrator PLUS for automated ISE techniques and titrations
- RS-232 interface for easy communication
Includes:
AT5050: 48-position beaker rack for 50mL beakers; small electrode holder; drip tray; cable management system; BNC extension cable; stirrer extension cable; accessories package; interface cable; line cord; 50 pack of 50mL plastic beakers; instruction manual; warranty card.
AT5150: 28-position beaker rack for 150mL beakers; small electrode holder; drip tray; cable management system; BNC extension cable; stirrer extension cable; accessories package; interface cable; line cord; 50 pack of 150mL plastic beakers; instruction manual; warranty card.
Compatible with:
- 1112000 - 3 Star Plus pH meter
- 1114000 - 3 Star Plus conductivity meter
- 1115000 - 4 Star Plus pH/ISE meter
- 1117000 - 4 Star Plus pH/conductivity meter
- 1119000 - 5 Star Plus pH/ISE/Conductivity/Dissolved Oxygen meter
- ATB050 - Beakers, 50mL, 50/pk
- ATB150 - Beakers, 150mL, 50/pk
- 096S22 - Beakers, 45mL, 50/pk
Specifications
Specifications
| Container Size | 150mL |
| Description | Autosampler with rack for 28 x 150mL beakers |
| Certifications/Compliance | CE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.