Learn More
Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes
Catalog No. 13301108
Change view
Click to view available options
For Use With (Equipment):
Orion 810 Series DO Probes
Orion 830A Series DO Probes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-301-108 Orion 810 Series DO Probes
13-640-18 Orion 830A Series DO Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-301-108 Supplier Thermo Scientific™ Supplier No. 080160
Add to Cart
Add to Cart
Help ensure proper and optimal probe performance with the Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes.
Help ensure proper and optimal probe performance with Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes. Adapters include stir funnel and electrode adapter sleeve for BOD bottle.
Warranty: Out of box.
TRUSTED_SUSTAINABILITY
Specifications
Description BOD Adapter
For Use With (Equipment) Orion 810 Series DO Probes
Type BOD Adapter
Includes Stir funnel and electrode adaptor sleeve
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.