Learn More
Thermo Scientific™ Orion™ Calcium Ion Selective Electrodes
Catalog No. 13641803
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
None
For Use With (Equipment):
Combination Calcium Electrode
Half-Cell Calcium Electrode
ISE Meter, Module 972001
Type:
Combination Calcium Electrode
Replacement Module
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types For Use With (Equipment) Type
13-641-803 None Half-Cell Calcium Electrode Replacement Module
13-301-5 None Combination Calcium Electrode Replacement Module
13-642-272 BNC (Waterproof) ISE Meter, Module 972001 Combination Calcium Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-641-803 Supplier Thermo Scientific™ Supplier No. 932001
Add to Cart
Add to Cart
Measure calcium ions in aqueous solutions simply, accurately and economically.
Measure calcium in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Calcium Electrodes.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small samples.
Related Products:
  • Calibration standards: 922006 (0.1 M CaCl2); 923206 (100ppm)
  • Required ISA: 932011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types None
Description Replacement Module for Half-Cell Calcium Electrode 9320BN
For Use With (Equipment) Half-Cell Calcium Electrode
Type Replacement Module