Learn More
Thermo Scientific™ Orion™ Chlorine Electrodes
Measure total residual chlorine quickly and accurately with these Chlorine electrodes.
Manufacturer:  Thermo Scientific™ 9770BNWP
 View more versions of this product
Catalog No. 13-642-274
Add to cart
Description
Description
Measure total residual chlorine quickly and accurately with the Thermo Scientific™ Orion™ Chlorine Electrodes. All forms of total residual chlorine are measured.
The method permits rapid analysis to be done at the sampling site, eliminating erroneous low readings due to Cl2 loss
  • Method requires addition of two reagents to the sample before measurement
  • Electrode has a slope of about +29mV per decade increase in concentration
  • Calibration standards catalog number: 100ppm as Cl2 (977007)
  • Required ISA catalog number: 977010 iodide reagent; 977011 acid reagent
Specifications
Specifications
Combination Ion Selective Electrode
Waterproof BNC
Waterproof BNC connector
ISE Meters
Documents
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.