Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Conductivity Calibration Standards

Catalog No. 1330178 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging:
Bottle
Bottles
Pouch
Conductivity:
100 μS/cm ± 2 μS/cm
111.9 mS/cm ± 0.8 mS/cm
12.9 mS/cm ± 0.1 mS/cm
12.9 mS/cm ± 0.1 mS/cm
1413 μS/cm ± 7 μS/cm
1413 μS/cm ± 7μS/cm
147 μS/cm ± 10 μS/cm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Conductivity
13-301-78 Bottle 100 μS/cm ± 2 μS/cm
13-301-77 Bottle 1413 μS/cm ± 7μS/cm
13-642-889 Bottles 111.9 mS/cm ± 0.8 mS/cm
09-357-10 Bottle 12.9 mS/cm ± 0.1 mS/cm
13-301-135 Bottle 12.9 mS/cm ± 0.1 mS/cm
13-642-887 Pouch 147 μS/cm ± 10 μS/cm
13-303-150 Pouch 111.9 mS/cm ± 0.8 mS/cm
13-642-888 Pouch 1413 μS/cm ± 7 μS/cm
13-642-890 Pouch 12.9 mS/cm ± 0.1 mS/cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-301-78 Supplier Thermo Scientific™ Supplier No. 011008
  • $277.50
    $219.65 / Pack of 5

    Save $57.85

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-accuracy conductivity calibration standards traceable to NIST™ standard reference materials, designed for reliable and reproducible measurements in lab and field. Available in a wide range of conductivity values to support consistent calibration, reliable measurements and regulatory.

Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.

Ensure accurate, reliable conductivity measurements with Thermo Scientific™ Orion™ Conductivity Calibration Standards, formulated for precision electrochemical analysis in both laboratory and field environments. These ready-to-use standards are traceable to NIST™ Standard Reference Materials (SRMs), supporting calibration accuracy, regulatory compliance and data reproducibility across a wide range of applications.

Key Features and Benefits

  • Traceable to NIST SRMs for accurate, compliant calibrations
  • High reproducibility for consistent results across labs and operators
  • Broad application use across QA/QC, environmental, industrial and research laboratories  
  • Optimized for Orion conductivity meters and electrode with integrated temperature compensation support

Designed for routine calibration and verification of conductivity meters and electrodes, these standards help reduce measurement variability and maintain consistent system performance. They are suitable for use in environmental monitoring, water and wastewater testing, food and beverage analysis and pharmaceutical quality control and manufacturing.

Complete Range of Conductivity Standards

Thermo Scientific Orion conductivity standards are available across a broad range of conductivity values to support everything from ultrapure water measurement to high-conductivity industrial samples. Common calibration points include:

  • 100 µS/cm – ideal for low conductivity and purified water applications
  • 147 µS/cm – for industrial calibration and method validation
  • 1413 µS/cm – general-purpose calibration for routine lab and field testing
  • 12.9 mS/cm (12,900 µS/cm) – suitable for higher conductivity samples
  • 111.9 mS/cm (111,900 µS/cm) – suitable for higher conductivity samples
  • 0.1 M KCl standards – for precise electrochemical calibration and method validation

Designed for compatibility with Thermo Scientific Orion conductivity meters and electrodes, including both 2-cell and Thermo Scientific™ DuraProbe™ 4-cell conductivity electrodes. Built-in temperature compensation tables in Orion meter software streamline calibrations by enabling automatic adjustments, reducing user error and saving time.

Pouch vs. Bottle Formats

Orion conductivity standards are available in 20 mL single-use pouches and 60 mL bottles to fit different workflows. Both formats deliver the same NIST-traceable accuracy and formulation consistency, enabling flexible use without compromising data quality.

  • Single-use pouches
    • Minimize contamination risk
    • Ensure fresh, accurate calibration every use
    • Ideal for field testing and regulated environments
    • Reduce handling and preparation time
  • Bottles
    • Cost-effective for high-throughput labs
    • Convenient for frequent calibrations
    • Suitable for controlled laboratory environments

With a long-standing reputation for quality and precision, Thermo Scientific Orion conductivity standards enable laboratories and field professionals to maintain measurement confidence, meet regulatory requirements and improve overall data quality. Whether performing routine calibration or critical analytical validation, these standards provide the accuracy and consistency essential for modern electrochemistry workflows.

TRUSTED_SUSTAINABILITY

Specifications

Description Conductivity Standard
Packaging Bottle
Certificate of Analysis Yes
Traceability to NIST Yes
Conductivity 100 μS/cm ± 2 μS/cm
Concentration 47 ppm as NaCl
Color Clear
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Electrodes
Solubility Soluble
Solution Type Standard Solution
Type Conductivity Calibration Standard
Quantity 5 x 60 mL
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.