Learn More
MSC Orion Cordage Pull Tape, 1/2 Inch, 3000 Foot, 1250 lb Strength, Clear
Catalog No. 5000536392
Encompass
This pull tape is a robust and reliable solution for various pulling applications, including cable pulling. With its clear design, it offers good visibility during use. It is engineered for professionals who require a durable and strong tape for demanding tasks.
  • High breaking strength of 1250 lbs.
  • Generous length of 3000 feet for extensive projects.
  • Clear tape material for enhanced visibility.
  • Durable construction ensures reliable performance.
  • Manufactured in Canada.
Catalog No. 50-005-36392 Supplier MSC Supplier No. 17233784
This pull tape is a robust and reliable solution for various pulling applications, including cable pulling. With its clear design, it offers good visibility during use. It is engineered for professionals who require a durable and strong tape for demanding tasks.
  • High breaking strength of 1250 lbs.
  • Generous length of 3000 feet for extensive projects.
  • Clear tape material for enhanced visibility.
  • Durable construction ensures reliable performance.
  • Manufactured in Canada.