Learn More
MSC Orion Cordage Spool Twine, Cotton, 1/16 inch Diameter, 20 lb Breaking Strength
Catalog No. 5000527120
Encompass
This spool twine is constructed from cotton and features a plied twist, making it suitable for various tying applications. It offers a breaking strength of 20 lb and a consistent diameter of 1/16 inch, providing reliable performance for general use.
  • Made from cotton material.
  • White color for versatile use.
  • Features a breaking strength of 20 lb.
  • Has a plied twist construction.
  • Offers 1,200 feet per pound.
  • Consistent diameter of 1/16 "
  • Packaged in a convenient coil container.
  • Total length of 1200 feet.
Catalog No. 50-005-27120 Supplier MSC Supplier No. 17233420
This spool twine is constructed from cotton and features a plied twist, making it suitable for various tying applications. It offers a breaking strength of 20 lb and a consistent diameter of 1/16 inch, providing reliable performance for general use.
  • Made from cotton material.
  • White color for versatile use.
  • Features a breaking strength of 20 lb.
  • Has a plied twist construction.
  • Offers 1,200 feet per pound.
  • Consistent diameter of 1/16 "
  • Packaged in a convenient coil container.
  • Total length of 1200 feet.