Learn More
Thermo Scientific™ Orion™ Cupric Ion Selective Electrodes
Catalog No. 1329988
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-299-88 BNC
13-645-157 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-299-88 Supplier Thermo Scientific™ Supplier No. 9429BN
Add to Cart
Add to Cart
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically.
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
  • Calibration Standards Catalog number: 0.1 M Cu(NO3)2/942906
  • Required ISA Catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description With BNC connector
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE