Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Double Junction Glass Bodied Combination pH Electrode PROMO

Catalog No. 13643100PM Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-643-100PM BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-643-100PM Supplier Thermo Scientific™ Supplier No. 9102DJWPPM
Get Up to 50% Off Electrochemistry Solutions
View all promotions
Add to Cart
Add to Cart

Capture accurate measurements of a variety of samples types, including solvents, with Orion glass body pH electrodes.

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.

  • 910008 3M KCl filling solution, 5 x 60 mL bottles
  • 910001 pH electrode storage solution, 475 mL bottle
  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475 mL, and 12 mm electrode storage bottle
  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
  • 910008 3M KCl filling solution, 60mL 1 bottle

Warranty period: 12 months from date of purchase

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC (Waterproof)
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type Double Junction pH Electrode
Filling Solution Type 3M KCl
Internal Reference Ag/AgCl Double Junction
Precision 0.02 pH
Storage Requirements Store in Orion pH storage solution 910001
Temperature (English) Operating 32°F to 140°F
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.