Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter

Catalog No. 13642789 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Meter, pH Electrode
pH/ISE Meter, ATC Probe
pH/ISE Meter, Ammonia ISE
pH/ISE Meter, Chloride ISE
pH/ISE Meter, Fluoride ISE
pH/ISE Meter, Nitrate ISE
pH/ISE Meter, Sodium ISE
pH/ISE Meter, pH Electrode
pH/ISE Meter, pH/ATC Electrode
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-642-789 pH/ISE Meter, Sodium ISE
13-642-782 Meter, pH Electrode
13-642-785 pH/ISE Meter, Ammonia ISE
13-642-786 pH/ISE Meter, Chloride ISE
13-642-787 pH/ISE Meter, Fluoride ISE
13-642-788 pH/ISE Meter, Nitrate ISE
13-642-781 pH/ISE Meter, pH Electrode
13-641-291 pH/ISE Meter, pH/ATC Electrode
13-642-780 pH/ISE Meter, ATC Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-642-789 Supplier Thermo Scientific™ Supplier No. 2115205

missing translation for 'validMainframeMsg'

Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with our pH/ISE benchtop meter for advanced lab analysis.

Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.

The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter's two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter's Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.

  • View large, individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
  • Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
  • EZ Startup menu facilitates quick and easy first-time meter setup
  • Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
  • Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
  • Non-volatile meter memory holds up to 1000 data points with time and date stamp
  • Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
  • Stir samples with ease using the meter-controlled Orion Star stirrer probe
  • Meter-attachable electrode stand assists with moving and maintaining electrodes
  • IP54-rated housing handles splashes and is wall-mountable to save space

Warranty: 3-year meter replacement warranty

Warranty and Services

3-year meter warranty

TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003)
Accuracy (ISE) ±0.2 mV or ±0.05% of reading whichever is greater
Outputs USB, RS232
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.1°C
Advanced ISE Functions Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit
Alarm Output High/low limit alarm
Backlight Option Yes, adjustable
Calibration pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (pH) 1 to 6
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 1000 with Date and Time Stamp
Description Orion Dual Star pH/ISE dual channel benchtop meter sodium & pH kit with ROSS sodium ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand
Calibration Points (ISE) 2 to 6
Channels 2
Measurement Modes pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature
Range (ISE) 0.0001 to 19,900 in ppm, M, mg/L, % or unit-less
Range (mV) ±1999.9 mV
Range (pH) -2.000 to 19.999
Resolution (ISE) 0.0001 minimum, 1 to 3 significant digits (user selectable)
Resolution (pH) 0.1, 0.01, 0.001
Temperature (Metric) Range -5.0°C to +105.0°C
Display Type Graphic LCD with Backlight
Includes Dual Star pH/ISE dual channel meter, 8611BNWP ROSS sodium ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 841108 Orion 1000 ppm sodium standard (475 mL), 841109 Orion KAP sodium standard & ISA solution (475 mL), 841111 Orion sodium ISA ionic strength adjuster (475 mL), 841113 Orion sodium electrode conditioning solution (475 mL), 841101 Orion sodium electrode storage solution (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
Inputs Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
Isopotential Point 7.000 pH (adjustable), 1.000 ISE (adjustable)
Keypad Numeric with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German
Log Function Type Automatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter
Power Supply Universal AC adapter (included)
Probe Type pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode, 927007MD Orion ATC probe)
Range (ORP) ±1999.9 mV
Range (Relative mV) ±1999.9 mV
Relative Humidity Range 5 to 85 %, non-condensing
Resolution (mV) 0.1 mV
Resolution (Temperature) 0.1°C, 0.1°F
Stability Smart Stability and Smart Averaging
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Selection Manual or automatic with ATC temperature probe
Type pH/ISE Meter, Sodium ISE
Warranty 3 Year Meter Warranty
Dimensions (L x W x H) 24 x 17 x 9 cm (9.5 x 6.7 x 3.5 in.)
Weight (English) 2 lb.
Weight (Metric) 0.9 kg
Electrical Requirements 90 to 260 VAC 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.