Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Catalog No. 1364149 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC
Screw Cap
Meter Input:
BNC
Phillips
Radiometer No. 7
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Meter Input
13-641-49 Screw Cap BNC
13-301-66 BNC Radiometer No. 7
13-641-53 Screw Cap Phillips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-641-49 Supplier Thermo Scientific™ Supplier No. 91CBNC
  • $208.50
    $185.65 / Each of 1
    Save $22.85
    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC inputs and electrodes with screw cap connectors
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Screw Cap
Description BNC; Screw Cap; Detachable Cable to BNC Connector
For Use With (Equipment) Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors
Type Electrode Adapter Cable
Meter Input BNC