Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Catalog No. p-4530066
Change view
Click to view available options
For Use With (Equipment):
Carbon Dioxide Electrode, 9502BNMP
HP Ammonia Electrode, 9512HPBNWP
HP Ammonia Electrode, 9512HPBNWP, 9512HP
Standard Ammonia Electrode, 9512BNWP, 9512
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-641-767 Carbon Dioxide Electrode, 9502BNMP
13-641-929 Standard Ammonia Electrode, 9512BNWP, 9512
13-641-931 Standard Ammonia Electrode, 9512BNWP, 9512
13-642-950 HP Ammonia Electrode, 9512HPBNWP, 9512HP
13-643-502 HP Ammonia Electrode, 9512HPBNWP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-641-767 Supplier Thermo Scientific™ Supplier No. 950204
Only null left
Add to Cart
Add to Cart
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
TRUSTED_SUSTAINABILITY
Specifications
Description For Carbon Dioxide Electrode 9502BNMP; Four membranes with O-rings
For Use With (Equipment) Carbon Dioxide Electrode, 9502BNMP
Type Gas-Sensing ISE Membrane
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.