Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ High Performance Ammonia Combination Ion Selective Electrode

Catalog No. 13643500 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-643-500 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-643-500 Supplier Thermo Scientific™ Supplier No. 9512HPBNWP
  • $1,820.00
    $1,399.65 / Each of 1

    Save $420.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtain quick, simple, accurate and economical results at low level ammonia measurements.

Obtain a more linear response at lower levels and improved response time in ammonia measurements with the Thermo Scientific™ Orion™ High-Performance Ammonia Electrode, an ammonia gas-sensing electrode.

Ammonia is an important aspect of wastewater and environmental management. The 9512HPBNWP complies with SM 4500-NH3 D-1997 or E-1997; ASTM D1426-08 (B).

Features:

  • 9512HPBNWP features a hydrophobic gas-permeable membrane technology for use with either loose membrane 951214 or pre-assembled membrane caps 951215. Good for samples with <0.1 mg/L ammonia.

Solutions required:

  • Standards
    • 951006 0.1 M NH4Cl standard, 475 mL bottle
    • 951007 1000 ppm ammonia as nitrogen standard, 475 mL bottle
    • 951207 100 ppm ammonia as nitrogen standard, 475 mL bottle
  • ISA
    • 951011 Alkaline reagent, 475 ml bottle
    • 951210 Ammonia low level ISA with blue dye, 475 mL bottle
    • 951211 Ammonia ISA with blue dye, 475 mL bottle
  • Electrode fill solution
    • 951209 Electrode filling solution, 60 mL bottle
  • Storage solution
    • 951213 Ammonia electrode storage solution, 475 mL bottle

Consumables required:

  • Membranes
    • 951214 Loose membranes for HP electrodes, box of 20
    • 951215 Pre-assembled outer body and membrane cap assembly for HP electrodes, 3 pack
  • Items included with
    • 951209 Electrode filling solution, 60mL 2 bottles
    • 951214 Loose membranes, box of 20. 1 box
    • 951215 Pre-assembled outer body and membrane cap assembly for HP electrodes, 3 pack

Warranty period:

  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY

Specifications

Applications Water testing, wastewater, biosolids, aquaculture, seafood, biological samples, wine, TKN, air.
Connector Types BNC (Waterproof)
Description Ammonia High Performance Ion Selective Electrode
Diameter (Metric) Body 12mm
Filling Solution Type High performance ammonia electrode filling solution
Internal Reference Volatile amines
Junction Annular PTFE
Length (Metric) 120mm
Length (Metric) Cable 1M
Material Epoxy
Temperature (English) Operating 32°F to 122°F
Certifications/Compliance ASTM D1426, Standard Methods 4500-NH3 D, EPA Method 1689, ISO 6778, OSHA ID-164.
For Use With (Equipment) ISE Meter with a BNC Connection
Temperature (Metric) Operating 0°C to 50°C
Type Ammonia Electrode
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.