Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
Catalog No. 1329991
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-299-91 BNC
13-645-160 BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-299-91 Supplier Thermo Scientific™ Supplier No. 9453BN
Add to Cart
Add to Cart
Get fast, stable and reliable iodide ions readings in aqueous solutions.
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
  • Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Description With BNC connector
For Use With (Equipment) ISE Meter
Type Solid state half-cell