Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
Catalog No. 13645160 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-645-160 BNC (Waterproof)
13-299-91 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-645-160 Supplier Thermo Scientific™ Supplier No. 9653BNWP
  • $1,944.00
    $1,677.65 / Each of 1
    Save $266.35
    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Get fast, stable and reliable iodide ions readings in aqueous solutions.
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
Features:
  • 9653BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Iodide Half-Cell Electrode 9453BN is to be used with the Orion 900200 sure flow double Junction electrode.
Solutions required:
  • Standard
    • 945306 0.1 M iodide standard, 475 mL bottle
  • ISA
    • 940011 ISA for iodide measurements, 475 mL bottle
  • Electrode fill solution
    • 900063 Optimum Results D electrode filling solution, 5 x 60 mL bottles
    • 900002 Inner fill solution for double junction ref electrode, 5 x 60 mL bottles
    • 900003 Outer fill solution for double junction ref electrode, 5 x 60 mL bottles
Items included with:
  • 9653BNWP
    • 900063 Optimum Results D electrode filling solution, 60 mL 1 bottle
    • 948201 Polishing strip. Qty 2.
    9453BN
    • 948201 Polishing strip. Qty 1.
Warranty period
  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC (Waterproof)
Description With waterproof BNC connector
Junction Sure-Flow Junction
Material Epoxy
For Use With (Equipment) ISE Meter
Temperature (Metric) Operating 0°C to 80°C
Type Combination ISE