Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Catalog No. 13641771 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
ISE Half Cell Electrodes
Ion Selective Electrode
Type:
Handle
Polishing Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
13-641-771 Ion Selective Electrode Polishing Strips
13-642-260 ISE Half Cell Electrodes Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-641-771 Supplier Thermo Scientific™ Supplier No. 948201
  • $247.00
    $184.65 / Pack of 24
    Save $62.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Description Polishing Strips for Solid State Ion Selective Electrodes
For Use With (Equipment) Ion Selective Electrode
Type Polishing Strips
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.