Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Ion Specific Electrode Kits

Catalog No. p-6956012
Change view
Click to view available options
Electrode:
Ammonia ISE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrode
13-301-143 Ammonia ISE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-301-143 Supplier Thermo Scientific™ Supplier No. 9512BNSLN

Ensure correct measurement by using high quality electrodes and reagents for ion specific measurement. Ordering is easy! All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis

Perform quick and accurate measurements by using high quality Thermo Scientific™ Orion™ Ion Specific Electrodes and reagents for ion specific measurement. Ordering is easy! We offer a comprehensive portfolio of ion selective electrode calibration standards, ionic strength adjusters, reagents, and fill solutions for your ISE measurement needs. All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis.

Fluoride ISE Kit

  • Orion Combination Fluoride ISE features the Thermo™ Scientific™ Orion™ Sure-Flow™ junction which contributes to highly stable and precise fluoride measurements while simplifying electrode maintenance.
  • 0.5, 1.0, 2.0, 5.0 and 10.0 ppm Fluoride Standards are pre-mixed with total ionic strength adjustor for your convenience. No dilution and no addition of ionic strength adjustor needed.
  • Total Ionic Strength Adjustor (TISAB II) included for addition to samples

Ammonia ISE Kit

  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
TRUSTED_SUSTAINABILITY

Specifications

Applications Ammonia Analysis
Connector Types BNC (Waterproof)
Description STD Ammonia Ion Selective Electrode and Reagent Kit
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12mm
Diameter (Metric) Cap 16mm
Filling Solution Type Std Ammonia ISE fill sol PN 951202
Internal Reference 9512BNWP
Junction Sure-Flow Junction
Length (English) 4.75 in.
Length (English) Cable 39.40 in.
Length (Metric) 150mm
Length (Metric) Cable 1m
Material Epoxy
Operating Range (Metric) Ranges : 1.0 to 5 x 10E-7M
Temperature (English) Operating 32-122°F
Membrane Ammonia Gas Sensing PN 951204
Response Time 1-4 min.
Model 9512BNWP
Electrode Ammonia ISE
Temperature (Metric) Range 0-50°C
For Use With (Equipment) ISE Meter
Temperature (Metric) Operating 25°C
Type Standard Ammonia ISE and Reagent Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.