Learn More
Description
Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).
Specifications
Specifications
| Chemical Name or Material | Sodium Lauryl Sulfate Standard |
| For Use With (Application) | PH ion conductivity and oxygen measurement |
| For Use With (Equipment) | Surfactant Ion Selective Electrode |
| Quantity | 60 mL |
| Solution Type | Surfactant Analysis |
| Packaging | Bottle |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.