Learn More
Thermo Scientific™ Orion™ Navigator Pro™ Computer Software
Description
Thermo Scientific™ Orion™ Navigator Pro™ 2.0 Computer Software and Thermo Scientific™ Orion™ Versa Star Pro™ meters are designed to offer a computer-driven measurement system for generating electronic measurements, calibration data and audit trails. Orion Navigator Pro software offers multi-level system access for an administrator and 100 users. The admin can customize each user's access level from full system access to basic data collection only. The Navigator Pro software controls all meter settings, data collection and reporting to help ensure data integrity and complete audit trail.
- Generates electronic records and electronic signatures
- Creates an audit trail report for electronic records
- Limits access to authorized individuals
- Password protection with system administrator and up to 100 users with customizable access levels
- Full computer control of meter functions including setup, calibration and measurement
- Paperless record keeping system
- Easily view and collect measurement data from your computer, including the ability to display up to four measurement channels on each Orion Versa Star Pro meter.
- Simplified setup menu allows instrument and measurement parameters to be set quickly and easily using your computer as the primary interface. Utilize your computer’s keyboard to enter sample IDs to improve sample throughput.
- Easy-to-view data records an be exported as a PDF file or sent to a network printer. Optional administrator-only configurable data output allows data records to be exported as PDF, Word, Excel or CSV files.
- Changes are tracked in the audit trail and can be saved as a PDF file for electronic records submissions and paperless record keeping.
Specifications
Specifications
| Connector Types | Requires one available USB 2.0, USB 3.0 or RS232 computer input |
| Description | Orion Navigator Pro 2.0 USB |
| Type | Computer Software |
| Includes | One license of computer software on USB flash drive with user manual |
| For Use With (Equipment) | Versa Star Pro Meters |