Learn More
Description
All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.
Buffer Color Codes:
- pH 4.01 buffer is pink
- pH 5.00 buffer is orange
- pH 7.00 buffer is yellow
- pH 10.01 buffer is blue
The buffers are available in pint (475 mL) bottles, portable five packs of 2 oz. (5 x 60 mL) bottles, convenient All-in-One buffer kits, 5 gallon (19 L) Cubitainers™ and single-use pouches.
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 10.01 ±0.02 |
| Physical Form | Liquid |
| Quantity | 475 mL |
| Solution Type | pH Buffer |
| Formulation | Water, Sodium Bicarbonate, Methylparaben, FD and C Blue #1, Sodium Carbonate |
| Packaging | Bottle |
| Traceability to NIST | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.