Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Buffer Cubitainers™

Catalog No. 1330171 Shop All Thermo Scientific Products
Change view
Click to view available options
pH:
4.01
7
10.01
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
13-301-71 10.01
13-301-69 4.01
13-301-70 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-301-71 Supplier Thermo Scientific™ Supplier No. 9110CB
  • $379.00
    $319.65 / Each of 1

    Save $59.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, 5 gallon cubitainers
Boiling Point 100°C
Color Light Blue
Certificate of Analysis Yes
Description pH 10.01 Buffer, 5 gallon
For Use With (Application) Accurate, reproducible pH calibrations
For Use With (Equipment) pH Meter and Electrode
pH 10.01
Physical Form Liquid
Quantity 19 L
Solution Type pH Buffer
Formulation Water, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
Packaging Cubitainer
Traceability to NIST Yes
Solubility Information Soluble
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.