Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Electrode Storage Solutions

Catalog No. 13303160
Change view
Click to view available options
For Use With (Equipment):
ROSS pH Electrode
pH Electrode
Quantity:
60 mL
475 mL
19 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
13-303-160 ROSS pH Electrode 475 mL
13-641-908 pH Electrode 475 mL
09-357-67 pH Electrode 60 mL
13-301-68 pH Electrode 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 13-303-160 Supplier Thermo Scientific™ Supplier No. 810001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.

Storage Solutions:

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples


Warranty: Out of box

TRUSTED_SUSTAINABILITY

Specifications

Description ROSS pH Electrode Storage Solution, 1 pint (475mL) Bottle
For Use With (Equipment) ROSS pH Electrode
Type pH Electrode Storage Solution
Filling Solution Type Storage
Packaging Bottle
Quantity 475 mL

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.