Learn More
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor Available on GSA/VA Contract for Federal Government customers only.
Sample well, rain, distilled/deionized, boiler feed and process waters with the Thermo Scientific? Orion? Pure Water? pH Buffers and pHISA? Adjustor.
Manufacturer:  Thermo Scientific™ 700001
 View more versions of this product
Catalog No. 13-641-735
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Features:
  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
Pure Water pH Test Kit:
  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
Description & Specifications
Specifications
pH Buffer/Ionic Strength Adjustor
provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
4 x 475mL pH 6.97 Buffer A, 2 x 475mL pH 4.10 Buffer B, 2 x 60mL pHISA Adjuster
475mL
Pure Water pH Test Kit
Kit
6.97
pH Meter and Electrode