Learn More
Thermo Scientific™ Orion™ RDO Calibration Sleeve
Catalog No. 13302129 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
RDO Probe
Catalog No. For Use With (Equipment)
13-302-129 RDO Probe
1 options
Catalog No. 13-302-129 Supplier Thermo Scientific™ Supplier No. 087003
  • $348.50
    $213.65 / Each of 1
    Save $134.85
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Replace a worn sleeve on an RDO probe with the Orion RDO Calibration Sleeve.
Replace the Thermo Scientific™ Orion™ RDO™ Calibration Sleeve on RDO probes as needed.
TRUSTED_SUSTAINABILITY
Specifications
Description RDO Calibration Sleeve
For Use With (Equipment) RDO Probe
Type Calibration Sleeve
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.