Learn More
Thermo Scientific™ Orion™ RDO Replacement Electrode Cap
Catalog No. 13302127 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Probe Optical Cap
Catalog No. Type
13-302-127 Probe Optical Cap
1 options
Catalog No. 13-302-127 Supplier Thermo Scientific™ Supplier No. 087001
  • $276.50
    $225.65 / Each of 1
    Save $50.85
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Use the probe optical cap replacement with RDO probes.
Use the Thermo Scientific™ Orion™ RDO™ Probe Optical Cap Replacement with Orion RDO probes.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Description Optical Cap
For Use With (Equipment) Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD
Type Probe Optical Cap
Do not use solvents or soaps to clean the RDO cap and do not rub the cap with abrasive material. Use of these materials will damage the RDO cap and void the warranty. Do not let salt or mineral deposits form on the RDO cap.