Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Catalog No. 13640
Change view
Click to view available options
Type:
Epoxy Refillable Triode
Epoxy Triode Low Maintenance
For Use With (Equipment):
Orion A+ Series Meters
Orion Star Series Meters
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
13-640 Epoxy Triode Low Maintenance Orion A+ Series Meters
13-642-255 Epoxy Triode Low Maintenance Orion Star Series Meters
13-642-256 Epoxy Refillable Triode Orion Star Series Meters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-640 Supplier Thermo Scientific™ Supplier No. 9179BN
Add to Cart
Add to Cart
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Features:
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • Built-in thermistor for Automatic Temperature Compensation (ATC)
  • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
TRUSTED_SUSTAINABILITY
Specifications
Description Epoxy Triode Low Maintenance
Material Epoxy
For Use With (Equipment) Orion A+ Series Meters
Type Epoxy Triode Low Maintenance