Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orion™ ROSS™ All-in-One™ pH Buffer Kit PROMO, Thermo Scientific™

Catalog No. 13641420PM Shop All Thermo Scientific Products
Change view
Click to view available options
ROSS All-in-One pH Buffer Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-641-420PM ROSS All-in-One pH Buffer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-641-420PM Supplier Thermo Scientific Supplier No. 810199PM

missing translation for 'validMainframeMsg'

Maintain pH electrodes more effectively with Thermo Scientific™ Orion™ ROSS™ All-in-One™ pH Buffer Kits.

Kit includes one each:

  • 1 pint (475mL) bottle of pH 4.01 buffer (910104)
  • 1 pint (475mL) bottle of pH 7.00 buffer (910107)
  • 1 pint (475mL) bottle of pH 10.01 buffer (910110)
  • 1 pint (475mL) bottle of ROSS™ pH electrode storage solution (810001)
  • 1 oz. (60 mL) bottle of general pH electrode cleaning solution (900024)
  • 12 mm diameter pH electrode storage bottle


Click here for the most complete, up-to-date MSDS and certificates of analysis files for Orion solutions

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ion Selective Electrode Calibration Standard
Description Fluoride Standard
For Use With (Equipment) Fluoride Ion Selective Electrode
Packaging Bottle

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.