Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ ROSS™ Combination Flat Surface pH Electrodes PROMO
Click to view available options
Connector Types:
BNC
Screw Cap
Description
The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.
- Material: Epoxy body
- pH range: 0 to 14
- Temperature range: 0 to 100°C
- Internal reference: ROSS
- Junction: Glass fiber
- Dimensions: 120 × 12 mm
- pH precision: 0.01
- Fill solution: Orion 810007
- Cap diameter: 16 mm
- Small samples are available in micro sample dish
- Flat surface tip
Includes: One 60 mL bottle of ROSS fill Solution, 810007 and spout cap

Specifications
Specifications
Description | With BNC connector |
Connector Types | BNC |
Junction | Glass Fiber |
Material | Epoxy Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Type | Combination Flat Surface pH Electrode, BNC Connector |
Drift | <0.002 pH/day |
Filling Solution Type | 3M KCl |
For Use With (Equipment) | Standard pH Meter |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction