Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS™ Sure-Flow™ pH Electrode with Sure-flow junction, BNC connector PROMO

Catalog No. 13642243PMO Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-642-243PMO BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-642-243PMO Supplier Thermo Scientific™ Supplier No. 8172BNWPPM

missing translation for 'validMainframeMsg'

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

This general purpose pH electrode offers superior performance for dirty, difficult samples such as soils, sludges, colloids, viscous materials and organics. Junction cleans easily between measurements; pressing down on the cap allows filling solution to flow through the junction carries away any contamination that may interfere with measurement.
  • Material: Glass body
  • pH range: 0 to 14
  • Temperature range: 0 to 100°C
  • Internal reference: ROSS
  • Junction: Sure-Flow
  • Dimensions: 120 × 12 mm
  • pH precision: 0.01
  • Fill solution: 3M KCl, Orion Cat. No 810007
  • Cap diameter: 16 mm

Includes:

  • Detachable bulb guard, two bottles of Orion ROSS electrode fill solution
TRUSTED_SUSTAINABILITY

Specifications

Description Orion ROSS Sure-Flow pH electrode
Connector Types BNC
Junction Sure-flow
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination pH Electrode, Glass Body, BNC Waterproof Connector
Filling Solution Type 3M KCl (810007)
For Use With (Application) Biological/Pharmaceutical –TRIS buffer, proteins, enzymes; Emulsions – Foods, cosmetics, oils; Emulsions – Petroleum products, paint; General Purpose – Most sample types; High Ionic Strength – Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength –
For Use With (Equipment) Standard pH Meter, BNC connector
Includes Detachable bulb guard, 2 x 60 mL bottles of ORion ROSS electrode fill solution
Internal Reference ROSS
Precision 0.01
Temperature (English) Operating 32 to 212°F
Diameter (Metric) Cap 16 mm
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.