Thermo Scientific™ Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes Available on GSA/VA Contract for Federal Government customers only.

No more waiting for pH readings to stabilize when measuring samples of varying temperatures.

Manufacturer: Thermo Scientific™ 8302BNUMD

 View more versions of this product

Catalog No. 13-641-780

  Check Availability
Add to Cart

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Warranty 2 years
    Description Orion ROSS Ultra Glass Triode pH/ATC Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
    Diameter (Metric) 12mm
    Length (Metric) Cable 1m
    Connector Types BNC Waterproof, 8-pin MiniDIN
    For Use With Thermo Scientific Orion Versa Star, Star A and Dual Star meters
    pH Range 0 to 14
    Internal Reference ROSS
    Precision 0.01
    pH Relative Accuracy 0.03pH for buffers using automatic temperature compensation
    Temperature (Metric) Operating 0° to 100°C
    Material Chemical resistant glass body
    Junction Ceramic
    Isopotential Point 7
    Filling Solution Type ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
    Length (Metric) 120mm

    No more waiting for pH readings to stabilize when measuring samples of varying temperatures. Our Thermo Scientific™ Orion™ ROSS Ultra™ Triode™ pH electrodes combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no Silver or Silver Chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.

    Measure pH and Temperature with One Electrode
    Complete your pH measurements faster. Our ROSS Ultra Triode pH electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.  Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters.
     
    Target Markets:
    • Pharmaceutical
    • Food Manufacturing
    • Academic/Government
    • Environmental
    Applications:
    • Quality Control
    • Research 
    • General Purpose

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit