Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes PROMO

Catalog No. 13641981PM2 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With: Orion Versa Star, Star A and Dual Star Meters, Connector: BNC Waterproof, 8-pin MiniDIN, Cable Length: 1 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-641-981PM2 For Use With: Orion Versa Star, Star A and Dual Star Meters, Connector: BNC Waterproof, 8-pin MiniDIN, Cable Length: 1 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-641-981PM2 Supplier Thermo Scientific™ Supplier No. 8107BNUMDPM

missing translation for 'validMainframeMsg'

Measure both pH and temperature in samples with these low maintenance, pH/ATC combination electrodes.

Increase the accuracy of your readings with the Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes. These combination electrodes have a pH sensor that delivers superior performance at varying sample temperatures. They combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no silver or silver chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.

Measure pH and Temperature with One Low-Maintenance Electrode
Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Features:

  • Made of Rugged Epoxy Body, Filled with Low Maintenance Gel
  • Connector Types: BNC Waterproof, 8-pin MiniDIN

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental

Applications:

  • Quality Control
  • Research
  • General Purpose
TRUSTED_SUSTAINABILITY
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.