Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrode, BNC Waterproof, 8-pin MiniDIN connector with 1m cable, 120 mL x 12 mm dia.; Waterproof BNC PROMO

Catalog No. 13643201PM Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-643-201PM BNC (Waterproof), 8-pin Mini-DIN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-643-201PM Supplier Thermo Scientific™ Supplier No. 8157BNUMDPM

missing translation for 'validMainframeMsg'

For any application when the use of a metallic reference electrode would contaminate the sample, you can rely on this combination electrode.

Unique internal element system replaces Ag/AgCl or Hg2Cl2 internals with platinum wire and redox-couple filling solution. Double-junction reference cells have porous ceramic or glass fiber outer junction. Design provides stable potential, independent of temperature, and stable readings even when samples vary by 50°C (90°F) or more. Minimizes drift, stirring effects, clogged junctions; no need for frequent standardization so samples are analyzed faster.

  • Epoxy body for durability and ruggedness
  • Built-in ATC probe
  • General-purpose applications
  • Standard size and standard bulb
  • Glass fiber junction
  • L × Dia.: 120 × 12 mm

Specifications

  • 0 to 14pH
  • 0° to 100°C (32° to 212°F) range
  • Slope, 92 to 102%; isopotential point, pH 7
  • When standardized in 6.86pH buffer at 25°C (77°F), accuracy is ±0.03pH for the buffer from 0° to 100°C (32° to 212°F) (using ATC); response is stable to 0.01pH within 30 seconds at 25°C (77°F) and 75°C (167°F) and stable to ±0.002pH within 15 seconds when cycling between pH 6.86 and 4.01 buffers at 25°C (77°F)
  • Max. cap dia. (top): 0.75 in.. (19 mm); collar dia. 0.62 in.. (16 mm); Cable length is 39.37 in.. (1m)
TRUSTED_SUSTAINABILITY

Specifications

Description Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and 8-pinminiDIN Connector and 1 meter cable
Connector Types BNC (Waterproof), 8-pin Mini-DIN
Junction Glass Fiber
Material Rugged Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH/ATC Combination Electrode
Filling Solution Type ROSS electrode 3 m KCl fill solution (Cat. No. 810007)
For Use With (Equipment) Orion Versa Star, Star A and Dual Starmeters
Internal Reference ROSS
Isopotential Point 7
pH Relative Accuracy 0.03 pH for buffers using automatic temperature compensation
Precision 0.01
Warranty 2 Years
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.