Learn More
Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set
Catalog No. 13645124 Shop All Thermo Scientific Products
Catalog No. 13-645-124 Supplier Thermo Scientific™ Supplier No. 1010004
  • $162.40
    $131.65 / Each of 1
    Save $30.75
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Connect your computer and printer to a Versa Star, Star A200 or Star A300 series meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set.
Connect your computer and printer to a meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set. These cables are designed for use with Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters.
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star series RS232 printer and computer cable set
Type Meter Interface Cable
Includes One RS232 printer interface cable (2.5mm RS232 meter connector and 25 pin RS232 printer connector) and one RS232 computer interface cable (2.5mm RS232 meter connector and 9 pin RS232 computer connector)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.