Learn More
Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter
Catalog No. 13645125 Shop All Thermo Scientific Products
Catalog No. 13-645-125 Supplier Thermo Scientific™ Supplier No. 1010005
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter.
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter. This adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star series RS232 to USB adapter cable
Type Meter Interface Cable
Includes RS232 to USB computer interface adapter and cable (2.5mm RS232 meter connector and USB computer connector)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.