Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probe Holder
Catalog No. 13645601 Shop All Thermo Scientific Products
Change view
Click to view available options
Conductivity and DO Probes Holder
Catalog No.
13-645-601 Conductivity and DO Probes Holder
1 options
This item is not returnable. View return policy
Catalog No. 13-645-601 Supplier Thermo Scientific™ Supplier No. STARAESCD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probes Holder with the Star A series portable meter armor to protect your sensors in the field
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probe Holder with the Star A series portable meter armor (Cat. No. STARA-AR) to protect, transport and store your sensors in the field. This conductivity and DO probe holder can be attached to either side of the portable meter armor and holds 12mm diameter or 15mm diameter conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve.
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star A series conductivity and dissolved oxygen sensor holder for portable meter protective armor
Type Meter Accessories
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.