Learn More
Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand
Catalog No. 13645597 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrode Arm
Catalog No.
13-645-597 Electrode Arm
1 options
Catalog No. 13-645-597 Supplier Thermo Scientific™ Supplier No. STARABEA
  • $348.50
    $291.65 / Each of 1
    Save $56.85
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage and minimize breakage
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage between samples and minimize electrode breakage using the stand's cable management system. Attach the stand to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters or use the stand with weighted base (Cat. No. STARA-HB) for compatibility with any meter system. Includes meter bracket, arm and electrode holder (Cat. No. STARA-ESCD).
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star A series meter stand, electrode holder and meter-attachable base
Type Electrode Stand/Holder
Includes Redesigned electrode holder (Cat .No. STARA-ELHD), plastic stand with cable management system and meter-attachable base
Recommended Accessories To use with almost any Orion benchtop or portable meter, purchase the weighted base separately (Cat. No. STARA-HB)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.