Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion Star™ A121 pH Portable Meter
SCI_ED

Catalog No. S04968 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Meter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S04968 Meter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04968 Supplier Thermo Scientific™ Supplier No. STARA1215/EMD

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

S04968 Meter Kit: Star A121 pH Meter with Batteries; 9107BNMD Gel-filled Epoxy pH/ATC Triode with 1.5 Meter Cable; 916099 pH Buffer 60mL Kit; 911110 Rinse Solution Pouches; STARA-CS Star A Meter Hard Carrying Case; STARA-AR Star A Meter Armor with Stand and Electrode Holder

Portable, waterproof meter provides simple operation and accurate readings every time.

Thermo Scientific Orion Star A121 pH Portable Meters combine simplicity with accuracy. A large LCD displays pH or mV readings along with temperature. Icons provide quick updates on battery life, electrode status and calibration information. Simple button layout and onscreen messages help with calibration and setup menu choices.

  • Large, informative screen clearly shows the main information, including pH or mV reading, temperature in °C or °F, meter mode, calibration information and battery life
  • Don't miss a reading - Auto-Read measurement mode locks in the stable reading on your screen and ready indicator alerts when readings are stable
  • Up to 3 point pH calibration
  • Automatic recognition of USA/NIST and DIN buffers
  • Easily recall calibration slope data for procedure checks
  • Non-volatile memory holds up to 50 data points
  • Four AA batteries (included) provide over 2000 hours of operation or purchase the universal power adapter (sold separately) to run meter on AC power
  • Perfectly portable, waterproof and protected to take anywhere with a IP67-rated housing
  • 3 year meter replacement warranty

Specifications

  • Accuracy (mV): ±0.2 mV or ±0.05% of reading whichever is greater
  • Accuracy (pH): ±0.01pH
  • Battery Life: 2000 hours
  • Inputs: BNC (pH), 8-pin Mini-DIN (temperature)
  • Probe Type: ±pH electrode, ATC probe, ORP electrode - purchase separately
  • Range (mV): 1600.0mV
  • Range (pH): -2.00 to 16.00pH
  • Relative Humidity Range: 5 to 85 %, non-condensing
  • Resolution (mV): 0.1mV
  • Resolution (pH): 0.1, 0.01pH
  • Temperature (Metric) Operating: 5 to 45°C
  • Temperature Range: -5° to +105°C
  • Calibration Points (pH): 1 to 3

Certifications

IP-67, CE, TV, FCC Class A limits

Warranty and Services

Three-year meter replacement warranty

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) pH, mV, Temperature
Temperature Range (Metric) -5°C to +105°C
Type Meter Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.