Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter

Catalog No. 13645611
Change view
Click to view available options
Type:
pH Meter
pH Meter Difficult Samples Kit
pH Meter Durable Sensor TRIS Kit
pH Meter Food/Beverage kit
pH Meter Separate Sensor Kit
pH Meter Standard Kit
pH Meter TRIS Kit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-645-611 pH Meter Durable Sensor TRIS Kit
13-645-519 pH Meter
13-303-201 pH Meter Difficult Samples Kit
13-303-202 pH Meter Standard Kit
13-303-203 pH Meter Separate Sensor Kit
13-303-204 pH Meter Food/Beverage kit
13-645-521 pH Meter TRIS Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-645-611 Supplier Thermo Scientific™ Supplier No. STARA2116

Reliably measure pH, mV, ORP and temperature with our benchtop pH meter for advanced lab analysis and printer or computer interfacing.

Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a 5 point pH calibration, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.

Thermo Scientific Orion Star A211 Benchtop pH Meters are easy-to-use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with universal power adapter or optional AA batteries
  • IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
  • Meters include CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY

Specifications

Description Orion Star A211 pH benchtop meter durable sensor TRIS kit with TRIS-compatible ROSS Ultra epoxy-body pH/ATC electrode
Accuracy (Temperature) ±0.1°C
Accuracy (pH) ±0.002
Data Management Details Transfer single, range or all data points to printer or computer
Outputs USB, RS-232
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Includes Star A211 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable
Measurement Modes pH, mV, Relative mV, ORP with Temperature
Calibration pH with calibration editing option, relative mV, ORP and temperature
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000.0
Range (ORP) ±2000.0mV
Resolution (mV) 0.1mV
Calibration Points 1 to 5
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 22.0°F to 221.0°F
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Temperature Probe Calibration 1 point temperature offset calibration
Display Type LCD, Graphic with Backlight
Backlight Option Yes (Selectable)
Memory Non-volatile memory preserves data log, calibration log and meter settings
Data Points 2000 with Date and Time Stamp
Log Function Type Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Alarm Output High/low limit alarm, calibration due alarm
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Dimensions (L x W x H) 24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
Weight (Metric) 0.9 kg
Weight (English) 2 lb.
Warranty 3 Year Meter Warranty
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Methods 10 per channel with password protection
Model Orion Star A211 pH/mV/RmV/ORP/Temperature Benchtop Meter
Power Supply Universal AC adapter (included) or 4 AA batteries (sold separately)
Probe Type pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode)
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean Available with Software Update
Range (Relative mV) ±2000.0mV
Relative Humidity Range 5 to 85%, non-condensing
Isopotential Point 7.000pH
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Channels 1
Battery Type 4 x AA (sold separately)
Type pH Meter Durable Sensor TRIS Kit
Battery Life 800 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.