Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Star A215 pH/conductivity Benchtop Multiparameter Meter Kit PROMO

Catalog No. 13645547PM
Change view
Click to view available options
Range (pH):
-2 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13-645-547PM -2 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-645-547PM Supplier Thermo Scientific™ Supplier No. STARA2155PM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Durable and dependable for a wide range of applications in any lab.

The Thermo Scientific Orion Star A215 pH/ Conductivity Benchtop Meter is the ideal choice for any lab. Get the information you need quickly and easily from the large, backlit graphic LCD display. Informative display shows individual pH/mV/RmV or conductivity/TDS/salinity/ resistivity results with temperature, or all results at once. Additional helpful information includes electrode status, time, date, sample ID, user ID and calibration. For advanced users, features such as stability and averaging choices provide additional options.
  • Anyone can use this meter with its plain language screen prompts, soft keys that update for easy selection and a multilanguage interface that supports English, Spanish, French, Italian, German and Chinese
  • Don't miss a reading — AUTO-READ™ locks in the stable reading on your screen, ready indicator alerts when readings are stable and timed reading gathers data in specific time intervals
  • Up to five point pH calibration with automatic recognition for USA/NIST and DIN buffers and calibration editing to fix errors without a complete recalibration
  • Selectable cell constant allows for use with 2- or 4-cell conductivity cells
  • For accurate conductivity results, selectable reading reference temperatures of 5°, 10°, 15°, 20° or 25°C with linear, nonlinear, nLFu or EP curve options
  • Choice of practical salinity or natural sea water curves for salinity readings
  • Linear or ISO/EN 27888 curves for TDS
  • Nonvolatile memory holds up to 2000 data points with time and date stamp
  • Easily transfer data and keep meter software up-to-date with the USB and RS232 ports and complimentary data analysis software
  • Mix samples without a stir plate with direct control of the Orion Star stirrer probe (096019, sold separately)
  • Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples
  • Works with almost every AC power source with the included universal power adapter or with four AA batteries (sold separately) to run the meter on DC power
  • IP54-rated housing handles splashes and is wall-mountable if table space is a problem
  • Solutions included: 14/13 conductivity standard, pH buffer and solution kit (10,7,4), storage solution and cleaning solution
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50 to 60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Outputs USB, RS232
Accuracy (Conductivity) 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Resistivity) 0.5% of reading ±1 digit
Accuracy (Salinity) 0.5% of reading ±1 digit
Accuracy (TDS) 0.5% of reading ±1 digit
Alarm Output High/low limit alarm, calibration due alarm
Backlight Option Yes (Selectable)
Battery Life 800 hr.
Battery Type 4 AA (sold separately)
Calibration pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (Conductivity) Up to 5
Calibration Points (pH) 1 to 5
Cell Constant 0.001 to 199.9 cm-1
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 2000 with Date and Time Stamp
Description Orion Star A215 pH/conductivity benchtop meter kit with ROSS Ultra Triode epoxy-body pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions and stand.
Channels 2
Display Type LCD Graphic, Backlit
IP Rating IP54
Measurement Modes pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature
Range (Conductivity) 0.001 μS/cm to 3000 mS/cm
Range (mV) ±2000.0 mV
Range (pH) -2 to 20
Range (Resistivity) 2 ohm to 100.0 meg-ohm
Range (Salinity) 0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS) 0.001 to 200.0 ppm
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
Isopotential Point 7 pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A215 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Benchtop Meter
Power Supply Universal AC adapter (included) or 4 AA batteries (sold separately)
Range (ORP) ±2000.0 mV
Range (Relative mV) ±2000.0 mV
Relative Humidity Range 5 to 85 %, non-condensing
Resolution (Conductivity) 0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (mV) 0.1 mV
Resolution (Resistivity) 1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity) 0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS) 0.001 ppm minimum, auto ranging up to 4 significant digits
Salinity Measurement Modes Practical salinity (psu) or natural sea water (ppt)
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
TDS Factor Range Linear (0.02 to 9.99)
Temperature Accuracy ±0.1°C
Temperature Compensation Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature (Metric) Reference 5, 10, 15, 20, 25°C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24 x 18 x 11 cm (9.5 x 7.1 x 4.3 in.)
Weight (English) 2 lb
Weight (Metric) 0.9 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.