Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion Star™ A221 Portable pH Meter PROMO

Catalog No. 13303210PM Shop All Thermo Scientific Products
Change view
Click to view available options
Range (pH):
-2 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13-303-210PM -2 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-303-210PM Supplier Thermo Scientific™ Supplier No. STARA2216PM

missing translation for 'validMainframeMsg'

Reliably measure pH, mV, ORP and temperature with our portable pH meter for advanced field analysis and printer or computer interfacing.

Don't leave your measurements to chance when analyzing samples outside the lab. Orion Star A221 Portable pH Meters are designed to offer the reliability you need in a user-friendly package for your dedicated pH testing needs. This waterproof, portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes – Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.2 mV or ±0.05%, whichever is greater
Accuracy (pH) ±0.002 pH
Data Management Details Transfer Single, Range or All Data Points to Printer or Computer
Outputs USB, RS-232
Includes Star A221 pH meter, 9107BNMD gel-filled epoxy-body pH/ATC electrode, pH 4/7/10 buffer and rinse pouches, storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
Measurement Modes pH, mV, Relative mV, ORP with Temperature
Calibration pH, Relative mV, ORP, Temperature
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000
Range (ORP) ±2000.0 mV
Resolution (mV) 0.1 mV
Calibration Points 1 to 5
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 22.0°F to 221.0°F
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Temperature Probe Calibration 1 point temperature offset calibration
Display Type LCD
Backlight Option Yes (Selectable)
Memory Non-volatile
Data Points 1000 with Date and Time Stamp
Log Function Type Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Alarm Output Calibration Due Alarm (Fixed 12 hour)
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP67
AC Adapter Universal 50/60 Hz, 100/240 VAC power adapter (sold separately, Cat. No. 1010003)
Dimensions (L x W x H) 24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (Metric) 0.5 kg
Weight (English) 1.2 lb.
Keypad Comprehensive with Menu-specific Function Keys and Dual Purpose Scroll/Shortcut Keys
Model Orion Star A221 pH/mV/RmV/ORP/Temperature Portable Meter
Power Supply 4 AA Batteries (included) or Universal AC Adapter (sold separately)
Probe Type pH Electrode, ORP Electrode, Reference Half-cell Electrode, ATC Temperature Probe (includes 9107BNMD silver chloride pH/ATC electrode)
Languages English, Spanish, French, Italian, German, Chinese, Portuguese and Korean available with software update
Range (Relative mV) ±2000.0 mV
Relative Humidity Range 5 to 85%, non-condensing
Isopotential Point 7 pH
Stability Smart Stability and Smart Averaging
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Channels 1
Battery Type 4 x AA
Type pH Meter Standard Kit
Battery Life 800 hours
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.