Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Star A325 pH, Conductivity and Temperature Portable Waterproof Multiparameter Meter Kit - PROMO

Catalog No. 13645565PM Shop All Thermo Scientific Products
Change view
Click to view available options
Range (pH):
-2 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13-645-565PM -2 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-645-565PM Supplier Thermo Scientific™ Supplier No. STARA3255PM

missing translation for 'validMainframeMsg'

Retrieve accurate results from any location with the Thermo ScientificTM Orion StarTM A325 pH/Conductivity Portable Multiparameter Meter.

Key Points:

  • Get the information you need quickly and easily from the large, backlit graphic LCD display, which shows individual pH/mV/RmV or conductivity/TDS/salinity/resistivity results with temperature, or all results at once.
  • Additional helpful information includes electrode status, time, date, sample ID, user ID and calibration points.
  • Advanced features such as stability and averaging provide additional options allowing for laboratory accuracy everywhere you need it.
  • Plain language screen prompts and soft keys that update for easy selection
  • Multi-language interface that supports English, Spanish, French, Italian, German and Chinese

Benefits:

  • Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen, ready indicator alerts when readings are stable and timed reading gathers data in specific time intervals
  • Up to five point pH calibration offers automatic recognition for USA/NIST™ and DIN buffers
  • Calibration editing allows user to fix pH calibration errors without a complete recalibration.
  • Selectable cell constant allows for use with 2- or 4-cell conductivity cells.
  • Selectable reading reference temperatures of 5°, 10°, 15°, 20° or 25°C with linear, nonlinear, nLFu or EP curve options offer advanced accuracy in conductivity readings.
  • User can choose practical salinity or natural sea water curves for salinity readings.
  • Linear or ISO/EN 27888 curves for TDS are available.
  • USB and RS232 ports and complimentary data analysis software allow user to easily transfer data and keep meter software up to date.
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50 to 60 Hz, 100 to 240 VAC power adapter (sold separately)
Outputs USB, RS232
Accuracy (Conductivity) 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Resistivity) 0.5% of reading ±1 digit
Accuracy (Salinity) 0.5% of reading ±1 digit
Accuracy (TDS) 0.5% of reading ±1 digit
Alarm Output High/low limit alarm, calibration due alarm
Backlight Option Yes (Selectable)
Battery Life 800 hr.
Battery Type 4 AA (included)
Calibration pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (Conductivity) Up to 5
Calibration Points (pH) 1 to 5
Cell Constant 0.001 to 199.9 cm-1
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 5000 with Date and Time Stamp
Description Orion Star A325 pH/conductivity portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case
Channels 2
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature
Range (Conductivity) 0.001 μS/cm to 3000 mS/cm
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 2 ohm to 100.0 meg-ohm
Range (Salinity) 0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS) 0.001 to 200.0 ppm
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Includes Orion STAR A325 pH/conductivity portable meter; 8107UWMMD Orion ROSS Triode pH/ATC gel-filled electrode with 3 m cable; 013010MD Orion DuraProbe conductivity cell with 3 m cable; 910410 pH 4.01 buffer pouches, pack of 10; 910710 pH 7.00 buffer pouches, pack of 10; 911010 pH 10.01 buffer pouches, pack of 10; 01100710 conductivity standard pouches, 1413 uS; 911110 rinse solution, pack of 10; protective armor with electrode holder; hard carrying case and batteries
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
Isopotential Point 7.000 pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A325 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter
Power Supply 4 AA batteries (included) or universal AC adapter (sold separately)
Range (ORP) ±2000 mV
Range (Relative mV) ±2000 mV
Relative Humidity Range 5 to 85%, non-condensing
Resolution (Conductivity) 0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (mV) 0.1 mV
Resolution (Resistivity) 1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity) 0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS) 0.001 ppm minimum, auto ranging up to 4 significant digits
Salinity Measurement Modes Practical salinity (psu) or natural sea water (ppt)
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
TDS Factor Range Linear (0.02 to 9.99)
Temperature Accuracy ±0.1°C
Temperature Compensation Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature (Metric) Reference 5, 10, 15, 20, 25°C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.