Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Star A329 pH, ISE, Conductivity, Dissolved Oxygen, RDO and Temperature Portable Waterproof Multiparameter Meter Kit - PROMO

Catalog No. 13645571PM Shop All Thermo Scientific Products
Change view
Click to view available options
Range (pH):
-2 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13-645-571PM -2 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-645-571PM Supplier Thermo Scientific™ Supplier No. STARA3295PM

missing translation for 'validMainframeMsg'

Get laboratory accuracy everywhere you need it with advanced meter features in this portable, IP67 waterproof meter.

Large, Backlit Informative Graphic LCD:

  • User can view each parameter individually, two parameters simultaneously or all three parameters on one screen; all options are shown with temperature.
  • Helpful icons are shown for electrode status, time, date, sample ID, user ID and calibration data.
  • Informative screen prompts guide meter setup and calibration procedures.
  • Menu-specific function keys automatically update to ensure easy meter navigation.
  • Multi-language interface supports English, Spanish, French, Italian, German and Chinese.
  • Thermo Scientific™ AUTO-READ™ locks in a stable reading on the screen; ready indicator alerts when continuous readings are stable and timed reading gathers data in specific time intervals.

pH, mV and ISE:

  • Up to five point pH calibration is offered with automatic recognition of USA/NIST™ and DIN buffers.
  • Up to five point ISE calibration with advanced features such as linear point-to-point, non-linear selectable auto-blank, and low-concentration range stability ensure accuracy whether ISE is done infrequently or with multiple samples every day
  • Calibration editing allows pH and ISE calibration errors to be corrected without a complete recalibration.

Conductivity:

  • Automatic conductivity measurement ranging based on sample conductance and calibrated cell constant for the ideal display resolution
  • Selectable reading reference temperatures of 5°, 10°, 15°, 20° or 25°C with linear, nonlinear, nLFu or EP curve options for advanced accuracy in conductivity readings
  • Choice of practical salinity or natural sea water curves for salinity readings

Dissolved Oxygen:

  • Automatically recognizes Orion polarographic and RDO optical/luminescence-based probes—no need to worry about compatibility
  • Provides easy, accurate DO results with automatic temperature, automatic/manual barometric pressure, and automatic salinity compensation
  • Calibrates by water-saturated air, air-saturated water, manual using a Winkler Titration or zero point calibration

Memory and Software:

  • Non-volatile memory holds up to 5,000 data points with time and date stamp.
  • USB and RS232 ports and complimentary Orion Star Com data transfer software make data transfer to a computer fast and simple.

Meter Includes:

  • Four AA batteries for more than 800 hours of operation, literature CD, printed quick start guide and USB computer cable
  • Three-year meter replacement warranty
  • Universal power adapter sold separately to use AC power
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50 to 60 Hz, 100 to 240 VAC power adapter (sold separately)
Outputs USB, RS232
Accuracy (Barometer) ±6 mmHg
Accuracy (Conductivity) 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (ISE) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Polarographic DO) ±0.2 mg/L or ±2% of reading, whichever is greater
Accuracy (RDO) ±0.1 mg/L up to 8 mg/L, ±0.2 mg/L from 8 to 20 mg/L, ±10% of reading up to 50 mg/L
Accuracy (Resistivity) 0.5% of reading ±1 digit
Accuracy (Salinity) 0.5% of reading ±1 digit
Accuracy (TDS) 0.5% of reading ±1 digit
Advanced ISE Functions Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit
Alarm Output High/low limit alarm, calibration due alarm
Backlight Option Yes (Selectable)
Barometric Pressure Correction 400 to 850 mmHg, automatic using built-in barometer or manual entry
Battery Life 800 hr.
Battery Type 4 AA (included)
Calibration pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
Calibration Modes Water-saturated air, air-saturated water, manual (Winkler titration), zero point
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (Conductivity) Up to 5
Calibration Points (ISE) 1 to 5
Calibration Points (pH) 1 to 5
Cell Constant 0.001 to 199.9 cm-1
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 5000 with Date and Time Stamp
Description Orion Star A329 pH/ISE/conductivity/RDO/DO portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, RDO optical DO sensor, solutions, meter armor and field case
Channels 3
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature
Percent Saturation Range (Polarographic DO) 0.0 to 600% saturation
Range (Conductivity) 0.001 μS/cm to 3000 mS/cm
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 2 ohm to 100.0 meg-ohm
Range (Salinity) 0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS) 0.001 to 200.0 ppm
Resolution (ISE) 0.0001 minimum, 1 to 3 significant digits (user selectable)
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Includes Star A329 pH/ISE/Conductivity/RDO/DO Meter with Batteries; 8107UWMMD ROSS Gel-filled Epoxy pH/ATC Triode with 3 Meter Cable; 013010MD DuraProbe Epoxy 4-Probe (K=0.475) Conductivity Probe with 3 Meter Cable; 087010MD RDO Optical DO Probe with 3 Meter Cable
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
Isopotential Point 7.000 pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A329 pH/ISE/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/RDO Optical/Dissolved Oxygen/Temperature Portable Meter
Percent Saturation Relative Accuracy (RDO) ±2 % saturation ≤ 200 % saturation, ±10 % saturation > 200 % saturation
Percent Saturation Resolution (Polarographic DO) 0.1, 1 % saturation
Percent Saturation Range (RDO) 0.0 to 500% saturation
Percent Saturation Relative Accuracy (Polarographic DO) ±2 % saturation or ±2% of reading whichever is greater
Percent Saturation Resolution (RDO) 0.1, 1 % saturation
Power Supply 4 AA batteries (included) or universal AC adapter (sold separately)
Pressure Range 400 to 850 mmHg
Pressure Resolution 1 mmHg
Range (ISE) 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none
Range (ORP) ±2000 mV
Range (Polarographic DO) 0.00 to 90 mg/L
Range (RDO) 0.00 to 50 mg/L
Range (Relative mV) ±2000 mV
Relative Humidity Range 5 to 85%, non-condensing
Resolution (Conductivity) 0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (mV) 0.1 mV
Resolution (Polarographic DO) 0.01, 0.1 mg/L
Resolution (RDO) 0.01, 0.1 mg/L
Resolution (Resistivity) 1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity) 0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS) 0.001 ppm minimum, auto ranging up to 4 significant digits
Salinity Factor 0.0 to 45.0 ppt, automatic correction using conductivity reading or manual entry of sample salinity
Salinity Measurement Modes Practical salinity (psu) or natural sea water (ppt)
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
TDS Factor Range Linear (0.02 to 9.99)
Temperature Accuracy ±0.1°C
Temperature Compensation Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature (Metric) Reference 5, 10, 15, 20, 25°C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.