Learn More
Thermo Scientific™ Orion™ 3-Cell Conductivity Electrode
Catalog No. 13644906 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC/2.5 mm phono tip
Catalog No. Connector Types
13-644-906 BNC/2.5 mm phono tip
1 options
Catalog No. 13-644-906 Supplier Thermo Scientific™ Supplier No. 014005
  • $341.00
    $300.65 / Each of 1
    Save $40.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Measure samples with the utmost versatility using this 3-electrode conductivity cell, which has high, standard and low conductivity measuring ranges.
Features
Rugged epoxy/stainless steel conductivity probe with removable guard to facilitate ease of cleaning.
Solutions required:
Standards
  • 011008 100 μS/cm conductivity/TDS standard, 5 x 60 mL bottles
  • 01100910 147 μS/cm conductivity standard, 10 pouches
  • 011007 1413 μS/cm conductivity/TDS standard, 5 x 60 mL bottles
  • 01100710 1413 μS/cm conductivity/TDS standard, 10 pouches
  • 011006 12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
  • 01100610 12.9 mS/cm conductivity/TDS standard, 10 pouches
  • 011005 111 mS/cm conductivity standard, 5 x 60 mL bottles
  • 01100510 111 mS/cm conductivity standard, 10 pouches
Warranty period
6 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 1
Type Conductivity Cell
Connector Types BNC/2.5 mm phono tip
Description Three-Electrode Conductivity Cell
Material Rugged Epoxy/Stainless Steel, Removable Guard
Max. Immersion 120 mm
Measurement Ranges 0 μS/cm to 200 mS/cm
Min. Immersion 25 mm