Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Total Alkalinity Test Kit

Catalog No. 13299110
Change view
Click to view available options
Quantity:
475 mL
4 x 475 mL
Type:
Alkalinitiy Reagent Refill
Alkalinity Standard/Control Refill
Total Alkalinity Test Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
13-299-110 475 mL Total Alkalinity Test Kit
13-299-111 4 x 475 mL Alkalinitiy Reagent Refill
13-299-112 475 mL Alkalinity Standard/Control Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-299-110 Supplier Thermo Scientific™ Supplier No. 700010
Add to Cart
Add to Cart

Save time by eliminating additional equipment set-up, titrations and calculations with this total alkalinity test kit that provides two-step direct measurement.

Save time by eliminating additional equipment set-up, titrations and calculations. The Thermo Scientific™ Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH using a pH electrode.

Features:

  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity

Includes:

  • One 1 pint (475mL) bottle alkalinity reagent (700011)
  • One 1 pint (475mL) bottle alkalinity standard/control (700012)
  • Total alkalinity conversion chart
  • Application procedure No. 517, Total Alkalinity in Natural Waters

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number: 700011

New Part Number: 700011-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Composition Non Hazardous Media
Format Liquid
For Use With (Application) two-step direct measurement of total alkalinity and pH using a pH electrode; Saves time by eliminating additional equipment set-up, tedious titrations and calculations
For Use With (Equipment) pH Electrode
Includes 475mL Alkalinity Reagent (700011), 475mL Alkalinity Standard/Control (700012), Total Alkalinity Conversion Chart and Application Procedure No. 517, Total Alkalinity in Natural Waters
Packaging Kit
Quantity 475 mL
Type Total Alkalinity Test Kit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.