Learn More
Thermo Scientific™ Orion™ Two-Electrode Conductivity Cells, Steel, V4A, Mini DIN connection, 0.01 to 300 μS/cm PROMO
Catalog No. 13642227PM Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
Mini-DIN
Catalog No. Connector Types
13-642-227PM Mini-DIN
1 options
Catalog No. 13-642-227PM Supplier Thermo Scientific™ Supplier No. 013016MDPM
Perform applications in either the lab or the field with these versatile two-electrode conductivity cells, available in a variety of materials and measurement ranges.
Perform applications in either the lab or the field with versatile Thermo Scientific™ Orion™ Two-Electrode Conductivity Cells, available in a variety of materials and measurement ranges.
2-Electrode conductivity cells 011010A and 011010 are glass/platinum, have a nominal cell constant of 1.0 cm-1 and are intended for laboratory use; measurement range is 1 μS/cm to 200 mS/cm.
2-Electrode conductivity cells 011020A, 011020 and 9902BN are glass/platinum, have a nominal cell constant of 0.1 cm-1 and are intended for laboratory use and ultra pure water; measurement range is 0.1 μS/cm to 100 μS/cm.
2-Electrode conductivity cells 011050MD, 011050A and 011050 are epoxy/platinum, have a nominal cell constant of 1.0 cm-1 and are intended for field or laboratory use; measurement range is 1 μS/cm to 20 mS/cm.
2-Electrode conductivity cells 011510MD and 011510are epoxy/graphite, have a nominal cell constant of 1.0 cm-1 and are intended for field or laboratory use; measurement range is 10 μS/cm to 200 mS/cm.
2-Electrode conductivity cells 013016MD, 013016A and 013016D are steel (V4A), have a nominal cell constant of 0.1 cm-1 and are intended for laboratory use and ultra pure water; measurement range is 0.01μS/cm to 300μS/cm
2-Electrode conductivity cell 018020MD is glass/platinum, has a nominal cell constant of 10 cm-1 and is intended for high electrolyte concentration solutions; measurement range is 10 μS/cm to 2000 mS/cm.
Recommended for:
Intended for laboratory and field use
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Mini-DIN
Type 2-Electrode Conductivity Cells
For Use With (Equipment) Orion Star Series Meters
Description 2-Electrode Conductivity Cell
Material Steel, V4A
Measurement Ranges 0.01 to 300 μS/cm